The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321136.1 | NP_001308065.1 | 13 - 14 | 1640 | 88 |
NM_001321137.1 | NP_001308066.1 | 13 - 14 | 1237 | 88 | |
NM_003978.4 | NP_003969.2 | 12 - 13 | 1421 | 88 | |
XM_006720737.2 | XP_006720800.1 | 12 - 13 | 1153 | 88 | |
XM_011522163.1 | XP_011520465.1 | 13 - 14 | 1439 | 88 | |
XM_011522164.1 | XP_011520466.1 | 13 - 14 | 1107 | 88 | |
XM_011522165.2 | XP_011520467.1 | 12 - 13 | 962 | 88 | |
XM_011522166.1 | XP_011520468.1 | 13 - 14 | 1473 | 88 | |
XM_011522168.2 | XP_011520470.1 | 13 - 14 | 1439 | 88 | |
GenBank mRNA | AB451310.1 | - | 12 - 13 | 932 | 88 |
AB451440.1 | - | 12 - 13 | 932 | 88 | |
AF038603.1 | - | 12 - 13 | 932 | 88 | |
AK308462.1 | - | 13 - 14 | 1271 | 88 | |
AK309856.1 | - | 13 - 14 | 1603 | 88 | |
BC008602.1 | - | 12 - 13 | 1293 | 88 | |
KU178458.1 | - | 12 - 13 | 932 | 88 | |
U94778.1 | - | 12 - 13 | 1148 | 88 |