The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320635.1 | NP_001307564.1 | 3 - 4 | 745 | 61 |
NM_001320636.1 | NP_001307565.1 | 3 - 4 | 377 | 61 | |
NM_020980.4 | NP_066190.2 | 3 - 4 | 745 | 61 | |
GenBank mRNA | AB008775.1 | - | 3 - 4 | 591 | 61 |
AF016495.1 | - | 3 - 4 | 603 | 61 | |
AK300055.1 | - | 3 - 4 | 350 | 61 | |
AK313214.1 | - | 3 - 4 | 670 | 61 | |
BC026258.1 | - | 3 - 4 | 560 | 61 | |
CR542027.1 | - | 3 - 4 | 375 | 61 | |
DC296768.1 | - | 3 - 4 | 381 | 61 |