The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317993.1 | NP_001304922.1 | 4 - 5 | 456 | 66 |
NM_001317994.1 | NP_001304923.1 | 3 - 4 | 973 | 66 | |
NM_018691.3 | NP_061161.2 | 3 - 4 | 409 | 66 | |
XM_005268360.4 | XP_005268417.1 | 4 - 5 | 534 | 66 | |
XM_005268361.4 | XP_005268418.1 | 3 - 4 | 437 | 66 | |
XM_011537540.2 | XP_011535842.1 | 3 - 4 | 763 | 66 | |
XM_011537541.2 | XP_011535843.1 | 3 - 4 | 1345 | 66 | |
XM_011537542.2 | XP_011535844.1 | 3 - 4 | 634 | 66 | |
XM_017008967.1 | XP_016864456.1 | 3 - 4 | 428 | 66 | |
XM_017008968.1 | XP_016864457.1 | 3 - 4 | 450 | 66 | |
GenBank mRNA | AF159700.1 | - | 3 - 4 | 902 | 66 |
AK024664.1 | - | 3 - 4 | 374 | 66 | |
AK225821.1 | - | 3 - 4 | 392 | 66 | |
AK313331.1 | - | 3 - 4 | 381 | 66 | |
BC011414.1 | - | 4 - 5 | 391 | 66 | |
BC050701.1 | - | 3 - 4 | 370 | 66 | |
DA351656.1 | - | 3 - 4 | 409 | 66 | |
DB022316.1 | - | 4 - 5 | 519 | 66 |