Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM114A2
Entrez Gene ID
10827
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
133K02, C5orf3
Chromosome Location
Chr.5: 153990131 - 154038938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317993.1 NP_001304922.1 5 - 6 542 151
NM_001317994.1 NP_001304923.1 4 - 5 1059 151
NM_001317995.1 NP_001304924.1 3 - 4 285 151
NM_018691.3 NP_061161.2 4 - 5 495 151
XM_005268360.4 XP_005268417.1 5 - 6 620 151
XM_005268361.4 XP_005268418.1 4 - 5 523 151
XM_011537540.2 XP_011535842.1 4 - 5 849 151
XM_011537541.2 XP_011535843.1 4 - 5 1431 151
XM_011537542.2 XP_011535844.1 4 - 5 720 151
XM_017008967.1 XP_016864456.1 4 - 5 514 151
XM_017008968.1 XP_016864457.1 4 - 5 536 151
GenBank mRNA AF159700.1 - 4 - 5 988 151
AK024664.1 - 4 - 5 460 151
AK225821.1 - 4 - 5 478 151
AK298943.1 - 3 - 4 254 151
AK303865.1 - 3 - 4 335 151
AK313331.1 - 4 - 5 467 151
BC011414.1 - 5 - 6 477 151
BC050701.1 - 4 - 5 456 151

View Full Product Details