The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317993.1 | NP_001304922.1 | 7 - 8 | 768 | 74 |
NM_001317994.1 | NP_001304923.1 | 6 - 7 | 1285 | 74 | |
NM_001317995.1 | NP_001304924.1 | 5 - 6 | 511 | 74 | |
NM_018691.3 | NP_061161.2 | 6 - 7 | 721 | 74 | |
XM_005268360.4 | XP_005268417.1 | 7 - 8 | 846 | 74 | |
XM_005268361.4 | XP_005268418.1 | 6 - 7 | 749 | 74 | |
XM_011537540.2 | XP_011535842.1 | 6 - 7 | 1075 | 74 | |
XM_011537541.2 | XP_011535843.1 | 6 - 7 | 1657 | 74 | |
XM_011537542.2 | XP_011535844.1 | 6 - 7 | 946 | 74 | |
XM_017008967.1 | XP_016864456.1 | 6 - 7 | 740 | 74 | |
XM_017008968.1 | XP_016864457.1 | 6 - 7 | 762 | 74 | |
GenBank mRNA | AF159700.1 | - | 6 - 7 | 1214 | 74 |
AK024664.1 | - | 6 - 7 | 686 | 74 | |
AK095446.1 | - | 6 - 7 | 641 | 74 | |
AK225821.1 | - | 6 - 7 | 704 | 74 | |
AK298943.1 | - | 5 - 6 | 480 | 74 | |
AK303865.1 | - | 5 - 6 | 561 | 74 | |
AK313331.1 | - | 6 - 7 | 693 | 74 | |
BC011414.1 | - | 7 - 8 | 703 | 74 | |
BC050701.1 | - | 6 - 7 | 682 | 74 |