Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM114A2
Entrez Gene ID
10827
Gene Name
family with sequence similarity 114 member A2
Gene Aliases
133K02, C5orf3
Chromosome Location
Chr.5: 153990131 - 154038938 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317993.1 NP_001304922.1 8 - 9 926 147
NM_001317994.1 NP_001304923.1 7 - 8 1443 147
NM_001317995.1 NP_001304924.1 6 - 7 669 147
NM_018691.3 NP_061161.2 7 - 8 879 147
XM_005268360.4 XP_005268417.1 8 - 9 1004 147
XM_005268361.4 XP_005268418.1 7 - 8 907 147
XM_011537540.2 XP_011535842.1 7 - 8 1233 147
XM_011537541.2 XP_011535843.1 7 - 8 1815 147
XM_011537542.2 XP_011535844.1 7 - 8 1104 147
XM_017008967.1 XP_016864456.1 7 - 8 898 147
XM_017008968.1 XP_016864457.1 7 - 8 920 147
GenBank mRNA AF159700.1 - 7 - 8 1372 147
AK024664.1 - 7 - 8 844 147
AK095446.1 - 7 - 8 799 147
AK225821.1 - 7 - 8 862 147
AK298943.1 - 6 - 7 638 147
AK303865.1 - 6 - 7 719 147
AK313331.1 - 7 - 8 851 147
BC011414.1 - 8 - 9 861 147
BC050701.1 - 7 - 8 839 147

View Full Product Details