The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317993.1 | NP_001304922.1 | 10 - 11 | 1141 | 86 |
NM_001317994.1 | NP_001304923.1 | 9 - 10 | 1658 | 86 | |
NM_001317995.1 | NP_001304924.1 | 8 - 9 | 884 | 86 | |
NM_018691.3 | NP_061161.2 | 9 - 10 | 1094 | 86 | |
XM_005268360.4 | XP_005268417.1 | 10 - 11 | 1219 | 86 | |
XM_005268361.4 | XP_005268418.1 | 9 - 10 | 1122 | 86 | |
XM_011537540.2 | XP_011535842.1 | 9 - 10 | 1448 | 86 | |
XM_011537541.2 | XP_011535843.1 | 9 - 10 | 2030 | 86 | |
XM_011537542.2 | XP_011535844.1 | 9 - 10 | 1319 | 86 | |
XM_017008967.1 | XP_016864456.1 | 9 - 10 | 1113 | 86 | |
XM_017008968.1 | XP_016864457.1 | 9 - 10 | 1135 | 86 | |
GenBank mRNA | AF159700.1 | - | 9 - 10 | 1587 | 86 |
AK024664.1 | - | 9 - 10 | 1059 | 86 | |
AK095446.1 | - | 9 - 10 | 1014 | 86 | |
AK225821.1 | - | 9 - 10 | 1077 | 86 | |
AK298943.1 | - | 8 - 9 | 853 | 86 | |
AK303865.1 | - | 8 - 9 | 934 | 86 | |
AK313331.1 | - | 9 - 10 | 1066 | 86 | |
BC011414.1 | - | 10 - 11 | 1076 | 86 | |
BC050701.1 | - | 9 - 10 | 1054 | 86 |