Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHX3
Entrez Gene ID
8022
Gene Name
LIM homeobox 3
Gene Aliases
CPHD3, LIM3, M2-LHX3
Chromosome Location
Chr.9: 136196250 - 136205129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014564.4 NP_055379.1 2 - 3 405 59
NM_178138.5 NP_835258.1 2 - 3 373 59
XM_005263410.1 XP_005263467.1 2 - 3 687 59
XM_017015168.1 XP_016870657.1 1 - 2 184 59
GenBank mRNA AB593062.1 - 2 - 3 373 59
AB593063.1 - 2 - 3 372 59
AB593064.1 - 2 - 3 687 59
AB593066.1 - 2 - 3 405 59
AB593067.1 - 2 - 3 369 59
AF096169.1 - 1 - 2 177 59
AF156888.1 - 2 - 3 360 59
AF156889.1 - 2 - 3 390 59

View Full Product Details