The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032932.5 | NP_116321.2 | 2 - 3 | 482 | 82 |
GenBank mRNA | AB058724.1 | - | 2 - 3 | 171 | 82 |
AJ314646.2 | - | 2 - 3 | 171 | 82 | |
AY169244.1 | - | 2 - 3 | 482 | 82 | |
BC093914.1 | - | 2 - 3 | 285 | 82 | |
BC101517.1 | - | 2 - 3 | 285 | 82 |