Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KATNAL1
Entrez Gene ID
84056
Gene Name
katanin catalytic subunit A1 like 1
Gene Aliases
-
Chromosome Location
Chr.13: 30202626 - 30307500 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014380.2 NP_001014402.1 2 - 3 291 122
NM_032116.4 NP_115492.1 2 - 3 333 122
XM_005266574.2 XP_005266631.1 2 - 3 475 122
XM_017020791.1 XP_016876280.1 2 - 3 346 122
XM_017020792.1 XP_016876281.1 2 - 3 367 122
GenBank mRNA AK097423.1 - 2 - 3 263 122
AK291439.1 - 2 - 3 266 122
AK315724.1 - 2 - 3 270 122
BC000612.2 - 2 - 3 293 122
DA085487.1 - 2 - 3 293 122
DR006891.1 - 2 - 3 368 122

View Full Product Details