The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014380.2 | NP_001014402.1 | 6 - 7 | 852 | 130 |
NM_032116.4 | NP_115492.1 | 6 - 7 | 894 | 130 | |
XM_005266574.2 | XP_005266631.1 | 6 - 7 | 1036 | 130 | |
XM_017020791.1 | XP_016876280.1 | 5 - 6 | 738 | 130 | |
XM_017020792.1 | XP_016876281.1 | 5 - 6 | 759 | 130 | |
XM_017020793.1 | XP_016876282.1 | 3 - 4 | 1068 | 130 | |
GenBank mRNA | AK097423.1 | - | 6 - 7 | 824 | 130 |
AK291439.1 | - | 6 - 7 | 827 | 130 | |
AK315724.1 | - | 6 - 7 | 831 | 130 | |
BC000612.2 | - | 6 - 7 | 854 | 130 |