Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGCA
Entrez Gene ID
6442
Gene Name
sarcoglycan alpha
Gene Aliases
50DAG, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin
Chromosome Location
Chr.17: 50165517 - 50175932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000023.3 NP_000014.1 7 - 8 1015 83
XM_011525120.2 XP_011523422.2 7 - 8 1481 83
XM_011525121.2 XP_011523423.2 7 - 8 1333 83
XM_011525124.2 XP_011523426.1 6 - 7 836 83
GenBank mRNA AK290622.1 - 7 - 8 988 83
BC025702.1 - 7 - 8 987 83
KU178331.1 - 7 - 8 959 83
L34355.1 - 7 - 8 996 83
U08895.1 - 7 - 8 970 83

View Full Product Details