The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014603.2 | NP_055418.2 | 3 - 4 | 754 | 70 |
XM_006721852.2 | XP_006721915.1 | 3 - 4 | 739 | 70 | |
GenBank mRNA | AK294006.1 | - | 3 - 4 | 754 | 70 |
BC047534.1 | - | 3 - 4 | 754 | 70 | |
L02867.1 | - | 3 - 4 | 552 | 70 |