Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF6
Entrez Gene ID
84295
Gene Name
PHD finger protein 6
Gene Aliases
BFLS, BORJ, CENP-31
Chromosome Location
Chr.X: 134373312 - 134428792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032335.3 NP_115711.2 5 - 6 620 80
GenBank mRNA AL832725.1 - 5 - 6 613 80
BC005994.1 - 5 - 6 559 80

View Full Product Details