The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021982.2 | NP_068817.1 | 14 - 15 | 2395 | 103 |
XM_006714523.2 | XP_006714586.1 | 13 - 14 | 2596 | 103 | |
XM_017008961.1 | XP_016864450.1 | 14 - 15 | 2040 | 103 | |
XM_017008962.1 | XP_016864451.1 | 14 - 15 | 1812 | 103 | |
GenBank mRNA | AJ131244.1 | - | 14 - 15 | 2058 | 103 |
AK304060.1 | - | 14 - 15 | 2384 | 103 | |
AL137677.1 | - | 6 - 7 | 674 | 103 |