The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252641.1 | NP_001239570.1 | 10 - 11 | 1422 | 99 |
XM_005259363.4 | XP_005259420.1 | 10 - 11 | 1473 | 99 | |
GenBank mRNA | BC026184.2 | - | 10 - 11 | 1419 | 99 |