The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040425.2 | NP_001035515.1 | 1 - 2 | 92 | 102 |
NM_144987.3 | NP_659424.2 | 1 - 2 | 92 | 102 | |
NR_110173.1 | 1 - 2 | 92 | 102 | ||
GenBank mRNA | AK094213.1 | - | 1 - 2 | 64 | 102 |
BM696851.1 | - | 1 - 2 | 85 | 102 | |
CD175846.1 | - | 1 - 2 | 101 | 102 |