The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110171.1 | 3 - 4 | 271 | 67 | |
NR_110172.1 | 3 - 4 | 271 | 67 | ||
NR_110173.1 | 3 - 4 | 296 | 67 | ||
GenBank mRNA | AY461582.1 | - | 3 - 4 | 245 | 67 |
BM696851.1 | - | 3 - 4 | 289 | 67 |