Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
Chromosome Location
Chr.16: 23835979 - 24220611 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002738.6 NP_002729.2 16 - 17 2060 97
GenBank mRNA BC036472.1 - 16 - 17 2020 97
X07109.1 - 16 - 17 1999 97

View Full Product Details