The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006612.5 | NP_006603.2 | 12 - 13 | 1372 | 68 |
XM_005256424.2 | XP_005256481.1 | 13 - 14 | 1363 | 68 | |
GenBank mRNA | AB014606.2 | - | 12 - 13 | 1368 | 68 |
BC034993.1 | - | 12 - 13 | 1346 | 68 | |
U91329.1 | - | 11 - 12 | 1128 | 68 |