Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF1C
Entrez Gene ID
10749
Gene Name
kinesin family member 1C
Gene Aliases
LTXS1, SATX2, SAX2, SPAX2, SPG58
Chromosome Location
Chr.17: 4997948 - 5028399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006612.5 NP_006603.2 14 - 15 1691 66
XM_005256424.2 XP_005256481.1 15 - 16 1682 66
GenBank mRNA AB014606.2 - 14 - 15 1687 66
AK307401.1 - 14 - 15 1534 66
BC034993.1 - 14 - 15 1665 66
U91329.1 - 13 - 14 1447 66

View Full Product Details