The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006612.5 | NP_006603.2 | 16 - 17 | 1843 | 63 |
XM_005256424.2 | XP_005256481.1 | 17 - 18 | 1834 | 63 | |
GenBank mRNA | AB014606.2 | - | 16 - 17 | 1839 | 63 |
BC034993.1 | - | 16 - 17 | 1817 | 63 | |
U91329.1 | - | 15 - 16 | 1599 | 63 |