The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003400.3 | NP_003391.1 | 18 - 19 | 2932 | 136 |
XM_005264544.1 | XP_005264601.1 | 17 - 18 | 2909 | 136 | |
XM_005264546.1 | XP_005264603.1 | 16 - 17 | 2040 | 136 | |
XM_006712094.2 | XP_006712157.1 | 19 - 20 | 3075 | 136 | |
XM_011533097.1 | XP_011531399.1 | 18 - 19 | 2241 | 136 | |
XM_011533098.1 | XP_011531400.1 | 17 - 18 | 2774 | 136 | |
XM_011533099.2 | XP_011531401.1 | 18 - 19 | 3882 | 136 | |
GenBank mRNA | AK124769.1 | - | 7 - 8 | 1809 | 136 |
AK289920.1 | - | 18 - 19 | 2611 | 136 | |
AK299039.1 | - | 9 - 10 | 1334 | 136 | |
AK312426.1 | - | 18 - 19 | 2369 | 136 | |
AK316481.1 | - | 9 - 10 | 1348 | 136 | |
AL833550.1 | - | 17 - 18 | 4877 | 136 | |
BC032847.2 | - | 19 - 20 | 2333 | 136 | |
BX647758.1 | - | 18 - 19 | 2914 | 136 | |
CR749840.1 | - | 19 - 20 | 2706 | 136 | |
D89729.1 | - | 17 - 18 | 2222 | 136 | |
Y08614.2 | - | 18 - 19 | 2242 | 136 |