Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XPO1
Entrez Gene ID
7514
Gene Name
exportin 1
Gene Aliases
CRM1, emb, exp1
Chromosome Location
Chr.2: 61477934 - 61538522 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003400.3 NP_003391.1 5 - 6 1084 108
XM_005264546.1 XP_005264603.1 3 - 4 192 108
XM_006712094.2 XP_006712157.1 6 - 7 1227 108
XM_011533097.1 XP_011531399.1 5 - 6 393 108
XM_011533098.1 XP_011531400.1 5 - 6 1061 108
XM_011533099.2 XP_011531401.1 5 - 6 2034 108
GenBank mRNA AK289920.1 - 5 - 6 763 108
AK312426.1 - 5 - 6 522 108
AL833550.1 - 6 - 7 2023 108
BC032847.2 - 6 - 7 485 108
BX647758.1 - 5 - 6 1066 108
CR749840.1 - 6 - 7 858 108
D89729.1 - 4 - 5 374 108
Y08614.2 - 5 - 6 394 108

View Full Product Details