The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289107.1 | NP_001276036.1 | 4 - 5 | 1337 | 61 |
NM_018414.4 | NP_060884.1 | 3 - 4 | 1200 | 61 | |
NR_110309.1 | 3 - 4 | 1200 | 61 | ||
XM_011524995.2 | XP_011523297.1 | 3 - 4 | 1120 | 61 | |
XM_011524996.2 | XP_011523298.1 | 3 - 4 | 1120 | 61 | |
XM_017024842.1 | XP_016880331.1 | 3 - 4 | 1120 | 61 | |
XM_017024843.1 | XP_016880332.1 | 3 - 4 | 1120 | 61 | |
XM_017024844.1 | XP_016880333.1 | 4 - 5 | 1220 | 61 | |
GenBank mRNA | AK000113.1 | - | 2 - 3 | 354 | 61 |
AY096001.1 | - | 3 - 4 | 1000 | 61 | |
AY358918.1 | - | 3 - 4 | 1145 | 61 | |
BC022462.1 | - | 3 - 4 | 1055 | 61 | |
Y11339.2 | - | 3 - 4 | 1060 | 61 |