Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTR1B
Entrez Gene ID
10120
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
Gene Aliases
ARP1B, CTRN2, PC3
Chromosome Location
Chr.2: 97655939 - 97664098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005735.3 NP_005726.1 7 - 8 959 94
XM_005263854.4 XP_005263911.1 6 - 7 718 94
XM_017003116.1 XP_016858605.1 7 - 8 1013 94
GenBank mRNA BC004374.2 - 7 - 8 827 94
BC006372.2 - 7 - 8 792 94
BC010090.2 - 7 - 8 859 94
BC025274.2 - 2 - 3 310 94
X82207.1 - 7 - 8 807 94
X82208.1 - 6 - 7 668 94

View Full Product Details