Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTR1B
Entrez Gene ID
10120
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
Gene Aliases
ARP1B, CTRN2, PC3
Chromosome Location
Chr.2: 97655939 - 97664098 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005735.3 NP_005726.1 5 - 6 648 82
XM_005263854.4 XP_005263911.1 4 - 5 407 82
XM_017003116.1 XP_016858605.1 5 - 6 702 82
GenBank mRNA BC004374.2 - 5 - 6 516 82
BC006372.2 - 5 - 6 481 82
BC010090.2 - 5 - 6 548 82
X82207.1 - 5 - 6 496 82
X82208.1 - 4 - 5 357 82

View Full Product Details