The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014141.5 | NP_054860.1 | 10 - 11 | 2190 | 79 |
XM_017011950.1 | XP_016867439.1 | 10 - 11 | 2516 | 79 | |
GenBank mRNA | AB020675.1 | - | 10 - 11 | 1699 | 79 |
AF319045.1 | - | 10 - 11 | 1814 | 79 | |
AK315113.1 | - | 10 - 11 | 1779 | 79 | |
BC093780.1 | - | 10 - 11 | 1717 | 79 | |
BC113373.1 | - | 10 - 11 | 1717 | 79 | |
KJ534811.1 | - | 9 - 10 | 1563 | 79 |