The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014141.5 | NP_054860.1 | 18 - 19 | 3527 | 85 |
GenBank mRNA | AB020675.1 | - | 18 - 19 | 3036 | 85 |
AB073883.1 | - | 6 - 7 | 947 | 85 | |
AF319045.1 | - | 18 - 19 | 3151 | 85 | |
AK294098.1 | - | 3 - 4 | 637 | 85 | |
AK296532.1 | - | 7 - 8 | 1183 | 85 | |
AK315113.1 | - | 18 - 19 | 3116 | 85 | |
BC093780.1 | - | 18 - 19 | 3054 | 85 | |
BC113373.1 | - | 18 - 19 | 3054 | 85 | |
KJ534810.1 | - | 2 - 3 | 554 | 85 | |
KJ534811.1 | - | 17 - 18 | 2900 | 85 | |
KJ534813.1 | - | 5 - 6 | 1010 | 85 | |
KJ535069.1 | - | - | 940 | 85 | |
KJ535088.1 | - | 7 - 8 | 1488 | 85 |