The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113755.2 | NP_001107227.1 | 8 - 9 | 1366 | 107 |
NM_001113756.2 | NP_001107228.1 | 7 - 8 | 1538 | 107 | |
NM_001257988.1 | NP_001244917.1 | 8 - 9 | 1389 | 107 | |
NM_001953.4 | NP_001944.1 | 8 - 9 | 1354 | 107 | |
GenBank mRNA | AK314716.1 | - | 8 - 9 | 1293 | 107 |
BC018160.1 | - | 8 - 9 | 1265 | 107 | |
BC052211.2 | - | 8 - 9 | 1303 | 107 | |
BF338223.1 | - | 2 - 3 | 284 | 107 | |
BM727471.1 | - | 2 - 3 | 364 | 107 | |
KU178037.1 | - | 7 - 8 | 1162 | 107 | |
M63193.1 | - | 7 - 8 | 1285 | 107 |