The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099691.2 | NP_001093161.1 | 1 - 2 | 288 | 98 |
NM_003236.3 | NP_003227.1 | 1 - 2 | 288 | 98 | |
GenBank mRNA | AK290151.1 | - | 1 - 2 | 99 | 98 |
AK312899.1 | - | 1 - 2 | 99 | 98 | |
BC005308.1 | - | 1 - 2 | 99 | 98 | |
DA216480.1 | - | 1 - 2 | 290 | 98 | |
DA462271.1 | - | 1 - 2 | 281 | 98 | |
K03222.1 | - | 1 - 2 | 74 | 98 | |
M31172.1 | - | 1 - 2 | 74 | 98 | |
X70340.1 | - | 1 - 2 | 71 | 98 |