Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PXDN
Entrez Gene ID
7837
Gene Name
peroxidasin
Gene Aliases
COPOA, D2S448, D2S448E, MG50, PRG2, PXN, VPO
Chromosome Location
Chr.2: 1631887 - 1744852 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012293.2 NP_036425.1 4 - 5 507 77
XM_011510396.1 XP_011508698.1 4 - 5 264 77
GenBank mRNA AF200348.1 - 4 - 5 466 77
BC098579.1 - 4 - 5 402 77
D86983.1 - 4 - 5 466 77
EF090903.1 - 4 - 5 466 77

View Full Product Details