Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIRT7
Entrez Gene ID
51547
Gene Name
sirtuin 7
Gene Aliases
SIR2L7
Chromosome Location
Chr.17: 81911939 - 81918182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016538.2 NP_057622.1 1 - 2 141 75
XM_011523580.1 XP_011521882.1 1 - 2 112 75
XM_011523581.2 XP_011521883.1 1 - 2 77 75
GenBank mRNA AF233395.1 - 1 - 2 123 75
AK002027.1 - 1 - 2 111 75
AK290265.1 - 1 - 2 109 75
BC017305.2 - 1 - 2 105 75
BC101791.1 - 1 - 2 122 75
BC101793.1 - 1 - 2 121 75
BQ890203.1 - 1 - 2 141 75

View Full Product Details