Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIRT7
Entrez Gene ID
51547
Gene Name
sirtuin 7
Gene Aliases
SIR2L7
Chromosome Location
Chr.17: 81911939 - 81918182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016538.2 NP_057622.1 4 - 5 459 87
XM_006722284.2 XP_006722347.1 2 - 3 193 87
XM_011523580.1 XP_011521882.1 3 - 4 325 87
XM_011523581.2 XP_011521883.1 3 - 4 2062 87
GenBank mRNA AF233395.1 - 4 - 5 441 87
AK002027.1 - 4 - 5 429 87
AK131437.1 - 1 - 2 1542 87
AK290265.1 - 4 - 5 427 87
BC017305.2 - 4 - 5 423 87
BC101791.1 - 4 - 5 440 87
BC101793.1 - 4 - 5 439 87
BQ890203.1 - 4 - 5 459 87
CD579273.2 - 4 - 5 445 87

View Full Product Details