The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243198.2 | NP_001230127.1 | 12 - 13 | 1168 | 99 |
NM_005338.6 | NP_005329.3 | 12 - 13 | 1168 | 99 | |
XM_005250304.2 | XP_005250361.1 | 12 - 13 | 1169 | 99 | |
XM_005250305.2 | XP_005250362.1 | 12 - 13 | 1174 | 99 | |
XM_011516116.2 | XP_011514418.1 | 12 - 13 | 1172 | 99 | |
XM_017012099.1 | XP_016867588.1 | 12 - 13 | 1089 | 99 | |
GenBank mRNA | AF365404.1 | - | 12 - 13 | 1095 | 99 |
AK302382.1 | - | 1 - 2 | 95 | 99 | |
BC110545.2 | - | 12 - 13 | 1143 | 99 | |
BC139786.1 | - | 2 - 3 | 222 | 99 | |
KF437291.1 | - | 12 - 13 | 1033 | 99 | |
U79734.1 | - | 11 - 12 | 991 | 99 | |
Y09420.1 | - | 4 - 5 | 384 | 99 |