The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243198.2 | NP_001230127.1 | 4 - 5 | 441 | 90 |
NM_005338.6 | NP_005329.3 | 4 - 5 | 441 | 90 | |
XM_005250304.2 | XP_005250361.1 | 4 - 5 | 442 | 90 | |
XM_005250305.2 | XP_005250362.1 | 4 - 5 | 447 | 90 | |
XM_011516116.2 | XP_011514418.1 | 4 - 5 | 445 | 90 | |
XM_017012099.1 | XP_016867588.1 | 4 - 5 | 362 | 90 | |
GenBank mRNA | AF365404.1 | - | 4 - 5 | 368 | 90 |
AK304738.1 | - | 4 - 5 | 410 | 90 | |
BC110545.2 | - | 4 - 5 | 416 | 90 | |
BP282107.1 | - | 4 - 5 | 434 | 90 | |
KF437291.1 | - | 4 - 5 | 306 | 90 | |
U79734.1 | - | 3 - 4 | 264 | 90 |