The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243198.2 | NP_001230127.1 | 9 - 10 | 855 | 60 |
NM_005338.6 | NP_005329.3 | 9 - 10 | 855 | 60 | |
XM_005250304.2 | XP_005250361.1 | 9 - 10 | 856 | 60 | |
XM_005250305.2 | XP_005250362.1 | 9 - 10 | 861 | 60 | |
XM_011516116.2 | XP_011514418.1 | 9 - 10 | 859 | 60 | |
XM_017012099.1 | XP_016867588.1 | 9 - 10 | 776 | 60 | |
GenBank mRNA | AF365404.1 | - | 9 - 10 | 782 | 60 |
AK304738.1 | - | 9 - 10 | 824 | 60 | |
BC110545.2 | - | 9 - 10 | 830 | 60 | |
KF437291.1 | - | 9 - 10 | 720 | 60 | |
U79734.1 | - | 8 - 9 | 678 | 60 | |
Y09420.1 | - | 1 - 2 | 71 | 60 |