Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATAD2B
Entrez Gene ID
57459
Gene Name
GATA zinc finger domain containing 2B
Gene Aliases
MRD18, P66beta, p68
Chromosome Location
Chr.1: 153804725 - 153922975 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005245364.4 XP_005245421.1 2 - 3 431 69
GenBank mRNA AB032976.1 - 1 - 2 56 69
AF411836.1 - 2 - 3 353 69
AK094665.1 - 2 - 3 580 69
BC069419.1 - 2 - 3 353 69
BC112052.1 - 2 - 3 357 69
BC112080.1 - 2 - 3 357 69

View Full Product Details