The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006371.4 | NP_006362.1 | 2 - 3 | 735 | 68 |
GenBank mRNA | AJ006470.1 | - | 2 - 3 | 626 | 68 |
AK001634.1 | - | 2 - 3 | 654 | 68 | |
AK091772.1 | - | 3 - 4 | 615 | 68 | |
AK092990.1 | - | 3 - 4 | 579 | 68 | |
AK126433.1 | - | 3 - 4 | 592 | 68 | |
AK298250.1 | - | 2 - 3 | 576 | 68 | |
AK314719.1 | - | 2 - 3 | 714 | 68 | |
BC008745.2 | - | 2 - 3 | 641 | 68 |