The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006371.4 | NP_006362.1 | 3 - 4 | 906 | 125 |
GenBank mRNA | AJ006470.1 | - | 3 - 4 | 797 | 125 |
AK001634.1 | - | 3 - 4 | 825 | 125 | |
AK075401.1 | - | 3 - 4 | 880 | 125 | |
AK091772.1 | - | 4 - 5 | 786 | 125 | |
AK092990.1 | - | 4 - 5 | 750 | 125 | |
AK126433.1 | - | 4 - 5 | 763 | 125 | |
AK298250.1 | - | 3 - 4 | 747 | 125 | |
AK314719.1 | - | 3 - 4 | 885 | 125 | |
BC008745.2 | - | 3 - 4 | 812 | 125 |