The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006371.4 | NP_006362.1 | 4 - 5 | 1045 | 120 |
GenBank mRNA | AJ006470.1 | - | 4 - 5 | 936 | 120 |
AK001634.1 | - | 4 - 5 | 964 | 120 | |
AK075401.1 | - | 4 - 5 | 1019 | 120 | |
AK091772.1 | - | 5 - 6 | 925 | 120 | |
AK126433.1 | - | 5 - 6 | 902 | 120 | |
AK298250.1 | - | 4 - 5 | 886 | 120 | |
AK314719.1 | - | 4 - 5 | 1024 | 120 | |
BC008745.2 | - | 4 - 5 | 951 | 120 |