The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015175.2 | NP_055990.1 | 32 - 33 | 5478 | 57 |
XM_006713072.3 | XP_006713135.1 | 31 - 32 | 5394 | 57 | |
XM_011533533.2 | XP_011531835.1 | 32 - 33 | 5475 | 57 | |
XM_017006010.1 | XP_016861499.1 | 32 - 33 | 5475 | 57 | |
XM_017006011.1 | XP_016861500.1 | 32 - 33 | 5331 | 57 | |
XM_017006012.1 | XP_016861501.1 | 31 - 32 | 5394 | 57 | |
XM_017006013.1 | XP_016861502.1 | 32 - 33 | 5475 | 57 | |
XM_017006014.1 | XP_016861503.1 | 31 - 32 | 5250 | 57 | |
XM_017006015.1 | XP_016861504.1 | 30 - 31 | 5106 | 57 | |
XM_017006016.1 | XP_016861505.1 | 27 - 28 | 6052 | 57 | |
XM_017006017.1 | XP_016861506.1 | 9 - 10 | 1894 | 57 | |
GenBank mRNA | AB011112.3 | - | 18 - 19 | 3162 | 57 |
AK131104.1 | - | 32 - 33 | 5318 | 57 | |
AY358455.1 | - | 2 - 3 | 428 | 57 |