Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH3
Entrez Gene ID
3699
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Gene Aliases
H3P, ITI-HC3, SHAP
Chromosome Location
Chr.3: 52794768 - 52809009 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002217.3 NP_002208.3 9 - 10 1118 85
XM_005265105.4 XP_005265162.1 9 - 10 1118 85
XM_017006356.1 XP_016861845.1 8 - 9 1024 85
GenBank mRNA AK222757.1 - 9 - 10 1090 85
AK223483.1 - 9 - 10 1116 85
AK298450.1 - 9 - 10 1090 85
BC107604.1 - 9 - 10 1088 85
BC107605.1 - 9 - 10 1088 85
BC107814.1 - 9 - 10 1088 85
X67055.1 - 9 - 10 1090 85

View Full Product Details