Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG4A
Entrez Gene ID
115201
Gene Name
autophagy related 4A cysteine peptidase
Gene Aliases
APG4A, AUTL2
Chromosome Location
Chr.X: 108088900 - 108154671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK054927.1 - 5 - 6 416 108

View Full Product Details