The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007237.2 | NP_001007238.1 | 3 - 4 | 1189 | 80 |
NM_001542.3 | NP_001533.2 | 2 - 3 | 1051 | 80 | |
XM_005270793.2 | XP_005270850.1 | 3 - 4 | 1161 | 80 | |
XM_005270794.4 | XP_005270851.1 | 2 - 3 | 459 | 80 | |
XM_006710593.3 | XP_006710656.1 | 2 - 3 | 462 | 80 | |
XM_011541315.1 | XP_011539617.1 | 3 - 4 | 1213 | 80 | |
GenBank mRNA | AB007935.2 | - | 2 - 3 | 521 | 80 |
AF031174.1 | - | 2 - 3 | 526 | 80 | |
BC110651.1 | - | 3 - 4 | 1189 | 80 | |
BC130279.1 | - | 2 - 3 | 859 | 80 | |
BC144132.1 | - | 2 - 3 | 859 | 80 |