The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 20 - 21 | 2539 | 80 |
NM_002209.2 | NP_002200.2 | 22 - 23 | 2791 | 80 | |
XM_005255313.1 | XP_005255370.1 | 22 - 23 | 2708 | 80 | |
XM_006721044.1 | XP_006721107.1 | 20 - 21 | 2493 | 80 | |
XM_011545849.1 | XP_011544151.1 | 22 - 23 | 2724 | 80 | |
GenBank mRNA | AK314246.1 | - | 22 - 23 | 2745 | 80 |
BC008777.2 | - | 20 - 21 | 2472 | 80 | |
Y00796.1 | - | 22 - 23 | 2703 | 80 |