The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015245.2 | NP_056060.2 | 15 - 16 | 2574 | 81 |
XM_005248964.3 | XP_005249021.1 | 16 - 17 | 2625 | 81 | |
XM_006715036.2 | XP_006715099.1 | 16 - 17 | 2625 | 81 | |
XM_011514431.2 | XP_011512733.1 | 16 - 17 | 2625 | 81 | |
XM_011514432.2 | XP_011512734.1 | 16 - 17 | 2625 | 81 | |
XM_011514433.1 | XP_011512735.1 | 16 - 17 | 2625 | 81 | |
XM_011514434.2 | XP_011512736.1 | 15 - 16 | 2562 | 81 | |
XM_011514435.2 | XP_011512737.1 | 15 - 16 | 2562 | 81 | |
XM_011514436.2 | XP_011512738.1 | 16 - 17 | 6194 | 81 | |
XM_011514437.2 | XP_011512739.1 | 6 - 7 | 2450 | 81 | |
XM_017010593.1 | XP_016866082.1 | 6 - 7 | 1051 | 81 | |
GenBank mRNA | BC031934.1 | - | 17 - 18 | 2240 | 81 |
BC049213.1 | - | 4 - 5 | 417 | 81 | |
BC132832.1 | - | 15 - 16 | 2511 | 81 | |
D86982.1 | - | 15 - 16 | 2571 | 81 |