The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017805.2 | NP_060275.2 | 9 - 10 | 2575 | 60 |
XM_011527053.2 | XP_011525355.2 | 9 - 10 | 2695 | 60 | |
XM_011527054.2 | XP_011525356.2 | 9 - 10 | 2695 | 60 | |
XM_011527055.2 | XP_011525357.2 | 6 - 7 | 2401 | 60 | |
XM_017026914.1 | XP_016882403.1 | 9 - 10 | 2695 | 60 | |
XM_017026915.1 | XP_016882404.1 | 6 - 7 | 2401 | 60 | |
GenBank mRNA | AK000408.1 | - | 5 - 6 | 690 | 60 |
AK123179.1 | - | 1 - 2 | 872 | 60 | |
AY378097.1 | - | 9 - 10 | 2440 | 60 | |
BC021860.1 | - | 5 - 6 | 705 | 60 | |
BC028614.1 | - | 9 - 10 | 2447 | 60 |