The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300946.1 | NP_001287875.1 | 2 - 3 | 412 | 73 |
NM_017660.3 | NP_060130.3 | 3 - 4 | 718 | 73 | |
XM_011528105.1 | XP_011526407.1 | 3 - 4 | 601 | 73 | |
XM_017026901.1 | XP_016882390.1 | 2 - 3 | 412 | 73 | |
XM_017026902.1 | XP_016882391.1 | 3 - 4 | 511 | 73 | |
XM_017026903.1 | XP_016882392.1 | 3 - 4 | 601 | 73 | |
XM_017026904.1 | XP_016882393.1 | 3 - 4 | 558 | 73 | |
XM_017026905.1 | XP_016882394.1 | 3 - 4 | 571 | 73 | |
XM_017026906.1 | XP_016882395.1 | 3 - 4 | 601 | 73 | |
XM_017026907.1 | XP_016882396.1 | 4 - 5 | 680 | 73 | |
XM_017026908.1 | XP_016882397.1 | 5 - 6 | 940 | 73 | |
XM_017026909.1 | XP_016882398.1 | 5 - 6 | 891 | 73 | |
XM_017026910.1 | XP_016882399.1 | 3 - 4 | 500 | 73 | |
GenBank mRNA | AK125974.1 | - | 5 - 6 | 823 | 73 |
AK315416.1 | - | 2 - 3 | 288 | 73 | |
AY186731.1 | - | 2 - 3 | 406 | 73 | |
BC011684.1 | - | 3 - 4 | 447 | 73 |