Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATAD2A
Entrez Gene ID
54815
Gene Name
GATA zinc finger domain containing 2A
Gene Aliases
p66alpha
Chromosome Location
Chr.19: 19385833 - 19508932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017026905.1 XP_016882394.1 1 - 2 161 106
GenBank mRNA AK303482.1 - 1 - 2 168 106

View Full Product Details